2-Methyl-5-(trifluoromethoxy)benzonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC303048-10G
PC303048-5G
PC303048-1G
APOSPC303048-10GEA
13160
CZK
APOSPC303048-10G
APOSPC303048-5G
APOSPC303048-1G
2-Methyl-5-(trifluoromethoxy)benzonitrile 97%
2-Methyl-5-(trifluoromethoxy)benzonitrile
Vzorec:
C₉H₆F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD18399750 CAS číslo: 1261679-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...