Tisk…

2-Methoxy-6-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

PC303043-1G PC303043-250MG
APOSPC303043-1GEA 7380 CZK
APOSPC303043-1G APOSPC303043-250MG
2-Methoxy-6-(trifluoromethyl)benzamide 97%
2-Methoxy-6-(trifluoromethyl)benzamide
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09832321
CAS číslo: 1017778-90-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR