Tisk…

2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)aniline

PC303034-5G PC303034-1G
APOSPC303034-5GEA 8700 CZK
APOSPC303034-5G APOSPC303034-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline
Vzorec: C₈H₇F₄NO
MW: 209,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201043
CAS číslo: 1262198-05-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR