2-Chloro-6-fluoro-3-methoxyphenol 98%
Dodavatel: Apollo Scientific
PC303031-10G
PC303031-5G
PC303031-25G
APOSPC303031-10GEA
11740
CZK
APOSPC303031-10G
APOSPC303031-5G
APOSPC303031-25G
2-Chloro-6-fluoro-3-methoxyphenol 98%
2-Chloro-6-fluoro-3-methoxyphenol
Vzorec:
C₇H₆ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09832266 CAS číslo: 1017777-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...