Tisk…

2-Chloro-5-(pentafluorothio)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Brommethyl)-1-chlor-4-(pentafluor-lambda6-sulphanyl)benzol, 2-Chloro-5-(pentafluorosulphur)benzyl bromide, [3-(Bromomethyl)-4-chlorophenyl]pentafluorosulphur

PC303029-1G PC303029-5G
APOSPC303029-1GEA 9530 CZK
APOSPC303029-1G APOSPC303029-5G
2-Chloro-5-(pentafluorothio)benzyl bromide 97%
2-Chloro-5-(pentafluorothio)benzyl bromide
Vzorec: C₇H₅BrClF₅S
MW: 331.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22988926
CAS číslo: 1431329-74-6
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR