2-Chloro-3-(trifluoromethyl)aniline 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC303026-5G
PC303026-25G
APOSPC303026-5GEA
622
CZK
APOSPC303026-5G
APOSPC303026-25G
2-Chloro-3-(trifluoromethyl)aniline 97%
2-Chloro-3-(trifluoromethyl)aniline
Vzorec:
C₇H₅ClF₃N Bod tání: 25…28 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115988 CAS číslo: 62476-58-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...