2,6-Difluoro-3-methoxybenzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC303023-5G
PC303023-25G
PC303023-50G
PC303023-100G
APOSPC303023-5GEA
4930
CZK
APOSPC303023-5G
APOSPC303023-25G
APOSPC303023-50G
APOSPC303023-100G
2,6-Difluoro-3-methoxybenzylamine 98%
2,6-Difluoro-3-methoxybenzylamine
Vzorec:
C₈H₉F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD04115905 CAS číslo: 886498-50-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...