2,6-Difluoro-3-methoxybenzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC303022-5G
PC303022-1G
PC303022-25G
APOSPC303022-5GEA
6390
CZK
APOSPC303022-5G
APOSPC303022-1G
APOSPC303022-25G
2,6-Difluoro-3-methoxybenzonitrile 98%
2,6-Difluoro-3-methoxybenzonitrile
Vzorec:
C₈H₅F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD04115902 CAS číslo: 886498-35-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...