2,4-Difluoro-3-methoxybenzonitrile 98%
Dodavatel: Apollo Scientific
PC303018-10G
PC303018-5G
PC303018-1G
APOSPC303018-10GEA
29760
CZK
APOSPC303018-10G
APOSPC303018-5G
APOSPC303018-1G
2,4-Difluoro-3-methoxybenzonitrile 98%
2,4-Difluoro-3-methoxybenzonitrile
Vzorec:
C₈H₅F₂NO MW: 169,13 g/mol Bod tání: 44…46 °C Storage Temperature: Ambient |
CAS číslo:
220353-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...