Tisk…

2,4-Difluoro-3-methoxyaniline 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Amino-2,6-difluoroanisole

PC303017-10G PC303017-5G PC303017-1G
APOSPC303017-10GEA 13460 CZK
APOSPC303017-10G APOSPC303017-5G APOSPC303017-1G
2,4-Difluoro-3-methoxyaniline 98%
2,4-Difluoro-3-methoxyaniline
Vzorec: C₇H₇F₂NO
MW: 159.14
Storage Temperature: Ambient
MDL Number: MFCD04115929
CAS číslo: 886499-08-7
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR