5-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC303005-1G
PC303005-5G
PC303005-10G
APOSPC303005-1GEA
5130
CZK
APOSPC303005-1G
APOSPC303005-5G
APOSPC303005-10G
5-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
5-Fluoro-2-(trifluoromethoxy)phenylacetic acid
Vzorec:
C₉H₆F₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD11519339 CAS číslo: 1092460-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...