5-Chloro-2-(trifluoromethoxy)benzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC303000-5G
PC303000-1G
APOSPC303000-5GEA
5200
CZK
APOSPC303000-5G
APOSPC303000-1G
5-Chloro-2-(trifluoromethoxy)benzoic acid 98%
5-Chloro-2-(trifluoromethoxy)benzoic acid
Vzorec:
C₈H₄ClF₃O₃ MW: 240,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519354 CAS číslo: 959749-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...