4-Methoxy-3-(trifluoromethoxy)benzoic acid 98%
Dodavatel: Apollo Scientific
PC302997-5G
PC302997-1G
APOSPC302997-5GEA
30780
CZK
APOSPC302997-5G
APOSPC302997-1G
4-Methoxy-3-(trifluoromethoxy)benzoic acid 98%
4-Methoxy-3-(trifluoromethoxy)benzoic acid
Vzorec:
C₉H₇F₃O₄ Storage Temperature: Ambient |
MDL Number:
MFCD18398451 CAS číslo: 647855-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...