4-Chloro-3,5-difluoroaniline 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC302981-1G
PC302981-5G
APOSPC302981-1GEA
1210
CZK
APOSPC302981-1G
APOSPC302981-5G
4-Chloro-3,5-difluoroaniline 97%
4-Chloro-3,5-difluoroaniline
Vzorec:
C₆H₄ClF₂N MW: 163,552 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09833604 CAS číslo: 2613-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...