4-Chloro-2-fluoro-3-methoxybenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302974-10G
PC302974-5G
PC302974-1G
APOSPC302974-10GEA
8500
CZK
APOSPC302974-10G
APOSPC302974-5G
APOSPC302974-1G
4-Chloro-2-fluoro-3-methoxybenzoic acid 98%
4-Chloro-2-fluoro-3-methoxybenzoic acid
Vzorec:
C₈H₆ClFO₃ MW: 204,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687204 CAS číslo: 1169870-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...