Tisk…

4-Chloro-2-(trifluoromethoxy)phenylacetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylacetic acid

PC302969-1G PC302969-5G
APOSPC302969-1GEA 5130 CZK
APOSPC302969-1G APOSPC302969-5G
4-Chloro-2-(trifluoromethoxy)phenylacetic acid 98%
4-Chloro-2-(trifluoromethoxy)phenylacetic acid
Vzorec: C₁₀H₈F₄O₂
MW: 254,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393873
CAS číslo: 1261570-91-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR