Tisk…

3-Methoxy-5-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

PC302961-10G PC302961-5G PC302961-1G
APOSPC302961-10GEA 11540 CZK
APOSPC302961-10G APOSPC302961-5G APOSPC302961-1G
3-Methoxy-5-(trifluoromethyl)benzamide 97%
3-Methoxy-5-(trifluoromethyl)benzamide
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09025411
CAS číslo: 916420-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR