Tisk…

3-Fluoro-5-(trifluoromethoxy)benzoic acid 98%

Dodavatel: Apollo Scientific

PC302957-1G PC302957-250MG
APOSPC302957-1GEA 7380 CZK
APOSPC302957-1G APOSPC302957-250MG
3-Fluoro-5-(trifluoromethoxy)benzoic acid 98%
3-Fluoro-5-(trifluoromethoxy)benzoic acid
Vzorec: C₈H₄F₄O₃
MW: 224,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783221
CAS číslo: 1242258-49-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR