Tisk…

3-Ethoxy-2,4-difluorobenzyl bromide 97%

Dodavatel: Apollo Scientific

PC302952-10G PC302952-5G PC302952-1G
APOSPC302952-10GEA 18190 CZK
APOSPC302952-10G APOSPC302952-5G APOSPC302952-1G
3-Ethoxy-2,4-difluorobenzyl bromide 97%
3-Ethoxy-2,4-difluorobenzyl bromide
Vzorec: C₉H₉BrF₂O
MW: 251,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258729
CAS číslo: 1017778-22-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR