Tisk…

3-Chloro-4-ethoxy-5-fluorobenzonitrile

Dodavatel: Apollo Scientific

PC302944-10G PC302944-5G
APOSPC302944-10GEA 11540 CZK
APOSPC302944-10G APOSPC302944-5G
3-Chloro-4-ethoxy-5-fluorobenzonitrile
3-Chloro-4-ethoxy-5-fluorobenzonitrile
Vzorec: C₉H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD09832345
CAS číslo: 1017778-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR