Tisk…

2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

PC302910-1G PC302910-5G PC302910-10G
APOSPC302910-1GEA 5030 CZK
APOSPC302910-1G APOSPC302910-5G APOSPC302910-10G
2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde 98%
2-Chloro-4,6-bis(trifluoromethyl)benzaldehyde
Vzorec: C₉H₃ClF₆O
Storage Temperature: Ambient
MDL Number: MFCD09025366
CAS číslo: 916420-48-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR