(5-Methyl-2-(trifluoromethyl)phenyl)acetic acid 97%
Dodavatel: Apollo Scientific
PC302896-1G
PC302896-5G
PC302896-10G
APOSPC302896-1GEA
4530
CZK
APOSPC302896-1G
APOSPC302896-5G
APOSPC302896-10G
(5-Methyl-2-(trifluoromethyl)phenyl)acetic acid 97%
(5-Methyl-2-(trifluoromethyl)phenyl)acetic acid
Vzorec:
C₁₀H₉F₃O₂ Bod tání: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832298 CAS číslo: 1017778-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...