Tisk…

5-Methyl-2-(trifluoromethyl)benzonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC302895-5G PC302895-1G
APOSPC302895-5GEA 4630 CZK
APOSPC302895-5G APOSPC302895-1G
5-Methyl-2-(trifluoromethyl)benzonitrile 97%
5-Methyl-2-(trifluoromethyl)benzonitrile
Vzorec: C₉H₆F₃N
MW: 185.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660290
CAS číslo: 886502-61-0
UN: 3276
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR