4-Methyl-3-(trifluoromethyl)benzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302889-10G
PC302889-5G
PC302889-1G
APOSPC302889-10GEA
14060
CZK
APOSPC302889-10G
APOSPC302889-5G
APOSPC302889-1G
4-Methyl-3-(trifluoromethyl)benzylamine 98%
4-Methyl-3-(trifluoromethyl)benzylamine
Vzorec:
C₉H₁₀F₃N MW: 189,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06212985 CAS číslo: 771581-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...