Tisk…

4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide

PC302880-5G PC302880-1G
APOSPC302880-5GEA 9660 CZK
APOSPC302880-5G APOSPC302880-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide
Vzorec: C₉H₆F₄NO
MW: 221,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201066
CAS číslo: 1373920-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR