Tisk…

3,5-Difluoro-4-ethoxybenzonitrile 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Ethoxy-3,5-difluorobenzonitrile

PC302875-5G PC302875-1G
APOSPC302875-5GEA 5130 CZK
APOSPC302875-5G APOSPC302875-1G
3,5-Difluoro-4-ethoxybenzonitrile 98%
3,5-Difluoro-4-ethoxybenzonitrile
Vzorec: C₉H₇F₂NO
MW: 183,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258687
CAS číslo: 1017779-27-9
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR