Tisk…

4-Chloro-3-ethoxy-2-fluorobenzylamine 98%

Dodavatel: Apollo Scientific

PC302867-1G PC302867-5G
APOSPC302867-1GEA 4530 CZK
APOSPC302867-1G APOSPC302867-5G
4-Chloro-3-ethoxy-2-fluorobenzylamine 98%
4-Chloro-3-ethoxy-2-fluorobenzylamine
Vzorec: C₉H₁₁ClFNO
Storage Temperature: Ambient
MDL Number: MFCD19687221
CAS číslo: 1323966-28-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR