Tisk…

4-Chloro-2-fluoro-3-methoxyaniline 98%

Dodavatel: Apollo Scientific
Varování

PC302860-1G PC302860-5G PC302860-10G
APOSPC302860-1GEA 4530 CZK
APOSPC302860-1G APOSPC302860-5G APOSPC302860-10G
4-Chloro-2-fluoro-3-methoxyaniline 98%
4-Chloro-2-fluoro-3-methoxyaniline
Vzorec: C₇H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD19687207
CAS číslo: 1323966-39-7
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR