3-Fluoro-5-(pentafluorothio)phenylacetonitrile 98%
Dodavatel: Apollo Scientific
PC302846-1G
PC302846-5G
APOSPC302846-1GEA
9920
CZK
APOSPC302846-1G
APOSPC302846-5G
3-Fluoro-5-(pentafluorothio)phenylacetonitrile 98%
3-Fluoro-5-(pentafluorothio)phenylacetonitrile
Vzorec:
C₈H₅F₆NS Storage Temperature: Ambient |
MDL Number:
MFCD16652460 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...