Tisk…

3-Fluoro-2-methoxybenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302842-1G PC302842-5G
APOSPC302842-1GEA 4530 CZK
APOSPC302842-1G APOSPC302842-5G
3-Fluoro-2-methoxybenzylamine 98%
3-Fluoro-2-methoxybenzylamine
Vzorec: C₈H₁₀FNO
MW: 155,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025368
CAS číslo: 916420-49-0
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR