Tisk…

3'-Chloro-5'-(pentafluorothio)acetophenone 97%

Dodavatel: Apollo Scientific

PC302833-1G PC302833-5G PC302833-10G
APOSPC302833-1GEA 9920 CZK
APOSPC302833-1G APOSPC302833-5G APOSPC302833-10G
3'-Chloro-5'-(pentafluorothio)acetophenone 97%
3'-Chloro-5'-(pentafluorothio)acetophenone
Vzorec: C₈H₆ClF₅OS
Storage Temperature: Ambient
MDL Number: MFCD16652455
CAS číslo: 1180486-96-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR