Tisk…

2-Methyl-5-(trifluoromethyl)benzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

PC302820-5G PC302820-1G PC302820-25G
APOSPC302820-5GEA 3740 CZK
APOSPC302820-5G APOSPC302820-1G APOSPC302820-25G
2-Methyl-5-(trifluoromethyl)benzaldehyde 98%
2-Methyl-5-(trifluoromethyl)benzaldehyde
Vzorec: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD06660200
CAS číslo: 886498-85-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR