Tisk…

2-Methyl-5-(trifluoromethoxy)benzoyl chloride 98%

Dodavatel: Apollo Scientific

PC302819-10G PC302819-5G PC302819-1G
APOSPC302819-10GEA 10250 CZK
APOSPC302819-10G APOSPC302819-5G APOSPC302819-1G
2-Methyl-5-(trifluoromethoxy)benzoyl chloride 98%
2-Methyl-5-(trifluoromethoxy)benzoyl chloride
Vzorec: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18399758
CAS číslo: 1261445-13-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR