2-Methoxy-5-(pentafluorothio)benzonitrile 98%
Dodavatel: Apollo Scientific
PC302812-1G
PC302812-5G
APOSPC302812-1GEA
9920
CZK
APOSPC302812-1G
APOSPC302812-5G
2-Methoxy-5-(pentafluorothio)benzonitrile 98%
2-Methoxy-5-(pentafluorothio)benzonitrile
Vzorec:
C₈H₆F₅NOS Storage Temperature: Ambient |
MDL Number:
MFCD16652491 CAS číslo: 1240256-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...