2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine
Dodavatel: Apollo Scientific
|
Varování
|
PC302807-100MG
PC302807-500MG
APOSPC302807-100MGEA
1370
CZK
APOSPC302807-100MG
APOSPC302807-500MG
2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine
2-Fluoro-5-methyl-4-trifluoromethyl-DL-phenylalanine
Vzorec:
C₁H₁F₄NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD19687203 CAS číslo: 1435806-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...