Tisk…

2-Fluoro-5-methyl-4-(trifluoromethyl)aniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-5-methyl-4-(trifluoromethyl)aniline

PC302806-1G PC302806-5G
APOSPC302806-1GEA 6620 CZK
APOSPC302806-1G APOSPC302806-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)aniline
2-Fluoro-5-methyl-4-(trifluoromethyl)aniline
Vzorec: C₈H₇F₄N
Storage Temperature: Refrigerator
MDL Number: MFCD22201049
CAS číslo: 1373920-66-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR