Tisk…

2-Chloro-6-fluoro-3-methoxybenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302796-10G PC302796-5G PC302796-1G
APOSPC302796-10GEA 10580 CZK
APOSPC302796-10G APOSPC302796-5G APOSPC302796-1G
2-Chloro-6-fluoro-3-methoxybenzylamine 98%
2-Chloro-6-fluoro-3-methoxybenzylamine
Vzorec: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD06660327
CAS číslo: 886761-60-0
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR