2,4-Difluoro-3-methoxybenzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302789-10G
PC302789-5G
PC302789-1G
APOSPC302789-10GEA
14060
CZK
APOSPC302789-10G
APOSPC302789-5G
APOSPC302789-1G
2,4-Difluoro-3-methoxybenzyl bromide
2,4-Difluoro-3-methoxybenzyl bromide
Vzorec:
C₈H₇BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD04115936 CAS číslo: 886499-17-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...