Tisk…

4-Chloro-2,6-difluorobenzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302778-5G PC302778-1G
APOSPC302778-5GEA 5460 CZK
APOSPC302778-5G APOSPC302778-1G
4-Chloro-2,6-difluorobenzylamine 98%
4-Chloro-2,6-difluorobenzylamine
Vzorec: C₇H₆ClF₂N
MW: 177,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660350
CAS číslo: 518357-43-2
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR