Tisk…

5-Methyl-2-(trifluoromethyl)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC302762-1G PC302762-5G PC302762-10G
APOSPC302762-1GEA 3870 CZK
APOSPC302762-1G APOSPC302762-5G APOSPC302762-10G
5-Methyl-2-(trifluoromethyl)phenylacetonitrile 97%
5-Methyl-2-(trifluoromethyl)phenylacetonitrile
Vzorec: C₁₀H₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD09832297
CAS číslo: 1017778-24-3
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR