Tisk…

4-Fluoro-3,5-dimethylbenzaldehyde 98%

Dodavatel: Apollo Scientific

PC302753-10G PC302753-5G PC302753-1G
APOSPC302753-10GEA 38350 CZK
APOSPC302753-10G APOSPC302753-5G APOSPC302753-1G
4-Fluoro-3,5-dimethylbenzaldehyde 98%
4-Fluoro-3,5-dimethylbenzaldehyde
Vzorec: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660264
CAS číslo: 363134-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR