Tisk…

4-Ethoxy-3,5-difluorobenzaldehyde 98%

Dodavatel: Apollo Scientific

PC302748-10G PC302748-5G PC302748-1G
APOSPC302748-10GEA 8040 CZK
APOSPC302748-10G APOSPC302748-5G APOSPC302748-1G
4-Ethoxy-3,5-difluorobenzaldehyde 98%
4-Ethoxy-3,5-difluorobenzaldehyde
Vzorec: C₉H₈F₂O₂
MW: 186.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247325
CAS číslo: 883536-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR