Tisk…

4'-Ethoxy-3',5'-difluoroacetophenone 98%

Dodavatel: Apollo Scientific

PC302747-10G PC302747-5G PC302747-1G
APOSPC302747-10GEA 8040 CZK
APOSPC302747-10G APOSPC302747-5G APOSPC302747-1G
4'-Ethoxy-3',5'-difluoroacetophenone 98%
4'-Ethoxy-3',5'-difluoroacetophenone
Vzorec: C₁₀H₁₀F₂O₂
MW: 200,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258688
CAS číslo: 1017779-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR