4-n-Butoxy-2,6-difluorophenylacetic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
4-Butoxy-2,6-difluorophenylacetic acid
PC302737-1G
PC302737-5G
APOSPC302737-1GEA
3870
CZK
APOSPC302737-1G
APOSPC302737-5G
4-n-Butoxy-2,6-difluorophenylacetic acid 98%
4-n-Butoxy-2,6-difluorophenylacetic acid
Vzorec:
C₁₂H₁₄F₂O₃ MW: 244,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201023 CAS číslo: 1373921-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...