4,5-Difluoro-2-methoxybenzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC302735-10G
PC302735-5G
PC302735-1G
APOSPC302735-10GEA
7010
CZK
APOSPC302735-10G
APOSPC302735-5G
APOSPC302735-1G
4,5-Difluoro-2-methoxybenzoic acid 97%
4,5-Difluoro-2-methoxybenzoic acid
Vzorec:
C₈H₆F₂O₃ MW: 188,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660196 CAS číslo: 425702-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...