Tisk…

3-Methoxy-2-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

PC302729-5G PC302729-1G PC302729-10G
APOSPC302729-5GEA 20500 CZK
APOSPC302729-5G APOSPC302729-1G APOSPC302729-10G
3-Methoxy-2-(trifluoromethyl)benzamide 97%
3-Methoxy-2-(trifluoromethyl)benzamide
Vzorec: C₉H₈F₃NO₂
MW: 219.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185678
CAS číslo: 1214351-75-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR