Tisk…

3-Methoxy-2-(trifluoromethyl)aniline 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-6-methoxybenzotrifluoride

PC302728-1G PC302728-250MG
APOSPC302728-1GEA 9690 CZK
APOSPC302728-1G APOSPC302728-250MG
3-Methoxy-2-(trifluoromethyl)aniline 98%
3-Methoxy-2-(trifluoromethyl)aniline
Vzorec: C₈H₈F₃NO
MW: 191,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185675
CAS číslo: 53982-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR