3-Fluoro-2-(trifluoromethyl)benzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302724-1G
PC302724-5G
PC302724-10G
APOSPC302724-1GEA
5130
CZK
APOSPC302724-1G
APOSPC302724-5G
APOSPC302724-10G
3-Fluoro-2-(trifluoromethyl)benzylamine 98%
3-Fluoro-2-(trifluoromethyl)benzylamine
Vzorec:
C₈H₇F₄N MW: 193,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06212983 CAS číslo: 771581-62-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...