3-Chloro-5-(trifluoromethoxy)benzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302716-5G
PC302716-1G
PC302716-25G
APOSPC302716-5GEA
3480
CZK
APOSPC302716-5G
APOSPC302716-1G
APOSPC302716-25G
3-Chloro-5-(trifluoromethoxy)benzaldehyde 98%
3-Chloro-5-(trifluoromethoxy)benzaldehyde
Vzorec:
C₈H₄ClF₃O₂ MW: 224,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660305 CAS číslo: 433926-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...