Tisk…

(3-Chloro-4-ethoxy-5-fluorophenyl)acetic acid 98%

Dodavatel: Apollo Scientific

PC302715-1G PC302715-5G PC302715-10G
APOSPC302715-1GEA 5130 CZK
APOSPC302715-1G APOSPC302715-5G APOSPC302715-10G
(3-Chloro-4-ethoxy-5-fluorophenyl)acetic acid 98%
(3-Chloro-4-ethoxy-5-fluorophenyl)acetic acid
Vzorec: C₉H₈ClFO₃
Storage Temperature: Ambient
CAS číslo: 1017778-96-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR